Gene Transcripts
- Gene Symbol
- PES1
- Entrez Gene ID
- 23481
- Gene Name
- pescadillo ribosomal biogenesis factor 1
- Gene Aliases
- PES
- Chromosome Location
- Chr. 22: 30576625 - 30607013 on Build GRCh38
- UniGene ID
- Hs.517543
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001243225.1 |
NP_001230154.1
|
4, 5 |
478 |
|
NM_001282327.1 |
NP_001269256.1
|
6, 7 |
876 |
|
NM_001282328.1 |
NP_001269257.1
|
6, 7 |
923 |
|
NM_014303.3 |
NP_055118.1
|
4, 5 |
478 |
|
XM_006724173.1 |
XP_006724236.1
|
4, 5 |
434 |
|
GenBank mRNA |
AB451380.1 |
- |
4, 5 |
371 |
|
AK096320.1 |
- |
6, 7 |
895 |
|
AK127185.1 |
- |
6, 7 |
877 |
|
AK315518.1 |
- |
4, 5 |
445 |
|
AU130886.1 |
- |
4, 5 |
445 |
|
BC032489.1 |
- |
4, 5 |
435 |
5416478 |
CR456539.1 |
- |
4, 5 |
396 |
|
U78310.1 |
- |
4, 5 |
429 |
|
View Full Product Details