Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025216.2 | NP_079492.2 | 3 | 990 | |
XM_011511929.2 | XP_011510231.1 | 4 | 598 | ||
GenBank mRNA | AB059569.1 | - | 3 | 532 | |
AK024363.1 | - | 3 | 990 | ||
AK315081.1 | - | 3 | 643 | ||
AY009400.1 | - | 3 | 723 | ||
BC003544.1 | - | 1 | 155 | ||
BC034352.1 | - | 3 | 649 | ||
BC040026.1 | - | 3 | 960 | ||
BC052234.1 | - | 3 | 1001 | 5442628 |