Gene Transcripts

Gene Symbol
USP51
Entrez Gene ID
158880
Gene Name
ubiquitin specific peptidase 51
Gene Aliases
Chromosome Location
Chr. X: 55484524 - 55489875 on Build GRCh38
UniGene ID
Hs.40061
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_201286.3 NP_958443.1 2 686
XM_017029299.1 XP_016884788.1 3 953
XM_017029300.1 XP_016884789.1 3 836
XM_017029301.1 XP_016884790.1 2 756
GenBank mRNA AJ583823.2 - 1 607
BC035907.1 - 2 697

View Full Product Details