Gene Transcripts
- Gene Symbol
- DNASE1
- Entrez Gene ID
- 1773
- Gene Name
- deoxyribonuclease I
- Gene Aliases
- DNL1, DRNI
- Chromosome Location
- Chr. 16: 3611705 - 3665472 on Build GRCh38
- UniGene ID
- Hs.629638
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_005223.3 |
NP_005214.2
|
3, 4 |
2343 |
|
XM_005255148.3 |
XP_005255205.1
|
6, 7 |
1254 |
|
XM_011522393.2 |
XP_011520695.1
|
6, 7 |
1253 |
|
XM_017022992.1 |
XP_016878481.1
|
6, 7 |
1255 |
|
XM_017022993.1 |
XP_016878482.1
|
7, 8 |
1860 |
|
XM_017022994.1 |
XP_016878483.1
|
8, 9 |
2026 |
|
XM_017022995.1 |
XP_016878484.1
|
6, 7 |
1060 |
|
XM_017022996.1 |
XP_016878485.1
|
6, 7 |
1006 |
|
XM_017022997.1 |
XP_016878486.1
|
7, 8 |
1674 |
|
XM_017022998.1 |
XP_016878487.1
|
5, 6 |
1959 |
|
XM_017022999.1 |
XP_016878488.1
|
4, 5 |
997 |
|
XM_017023003.1 |
XP_016878492.1
|
3, 4 |
1162 |
|
XM_017023007.1 |
XP_016878496.1
|
6, 7 |
1254 |
|
GenBank mRNA |
AB188151.1 |
- |
4, 5 |
422 |
|
AB188152.1 |
- |
3, 4 |
476 |
|
AJ298844.1 |
- |
3, 4 |
1298 |
|
BC029437.1 |
- |
3, 4 |
364 |
4594247 |
M55983.1 |
- |
3, 4 |
395 |
|
View Full Product Details