Gene Transcripts

Gene Symbol
TBC1D26
Entrez Gene ID
353149
Gene Name
TBC1 domain family member 26
Gene Aliases
Chromosome Location
Chr. 17: 15732277 - 15744784 on Build GRCh38
UniGene ID
Hs.434123
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178571.4 NP_848666.2 15 1400
GenBank mRNA BC028414.1 - 12 3638
BC047400.1 - 15 1394 5297645

View Full Product Details