Gene Transcripts
- Gene Symbol
- ELMOD1
- Entrez Gene ID
- 55531
- Gene Name
- ELMO domain containing 1
- Gene Aliases
- Chromosome Location
- Chr. 11: 107591091 - 107666779 on Build GRCh38
- UniGene ID
- Hs.495779
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001130037.1 |
NP_001123509.1
|
9, 10 |
1078 |
|
NM_001308018.1 |
NP_001294947.1
|
11, 12 |
1238 |
|
NM_018712.3 |
NP_061182.3
|
10, 11 |
1102 |
|
XM_017017994.1 |
XP_016873483.1
|
10, 11 |
1209 |
|
XM_017017995.1 |
XP_016873484.1
|
9, 10 |
959 |
|
GenBank mRNA |
AK054794.1 |
- |
9, 10 |
898 |
|
AK297351.1 |
- |
11, 12 |
1238 |
|
AK303689.1 |
- |
9, 10 |
920 |
|
AL359601.1 |
- |
4, 5 |
289 |
|
BC028725.2 |
- |
10, 11 |
845 |
4815682 |
View Full Product Details