Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001170690.1 | NP_001164161.1 | 5 | 610 | |
NM_020974.2 | NP_066025.2 | 5 | 610 | ||
XM_005253032.4 | XP_005253089.2 | 5 | 606 | ||
XM_005253033.4 | XP_005253090.2 | 5 | 606 | ||
XM_005253034.4 | XP_005253091.2 | 5 | 606 | ||
XM_005253035.4 | XP_005253092.2 | 5 | 606 | ||
XM_005253036.4 | XP_005253093.2 | 5 | 606 | ||
XM_005253037.4 | XP_005253094.2 | 5 | 606 | ||
XM_011520246.2 | XP_011518548.1 | 5 | 606 | ||
XM_011520247.2 | XP_011518549.1 | 5 | 606 | ||
XM_011520248.2 | XP_011518550.1 | 5 | 606 | ||
XM_017018080.1 | XP_016873569.1 | 5 | 606 | ||
XM_017018081.1 | XP_016873570.1 | 5 | 606 | ||
XM_017018082.1 | XP_016873571.1 | 5 | 606 | ||
GenBank mRNA | AK123039.1 | - | 5 | 606 | |
AK131552.1 | - | 5 | 604 | ||
BC111690.1 | - | 5 | 553 | 40008764 | |
DN995815.1 | - | 5 | 638 |