Gene Transcripts
- Gene Symbol
- SEL1L3
- Entrez Gene ID
- 23231
- Gene Name
- SEL1L family member 3
- Gene Aliases
- Sel-1L3
- Chromosome Location
- Chr. 4: 25697790 - 25863595 on Build GRCh38
- UniGene ID
- Hs.479384
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001297592.1 |
NP_001284521.1
|
13, 14 |
2203 |
|
NM_001297594.1 |
NP_001284523.1
|
13, 14 |
2225 |
|
NM_015187.4 |
NP_056002.2
|
13, 14 |
2420 |
|
XM_011513819.2 |
XP_011512121.2
|
13, 14 |
2414 |
|
XM_017007923.1 |
XP_016863412.1
|
12, 13 |
2058 |
|
GenBank mRNA |
AB018289.1 |
- |
12, 13 |
1918 |
|
AK025282.1 |
- |
1, 2 |
86 |
|
AK289687.1 |
- |
13, 14 |
2168 |
|
AK300219.1 |
- |
4, 5 |
518 |
|
BC009945.2 |
- |
6, 7 |
935 |
|
BC029422.1 |
- |
3, 4 |
345 |
|
BC041046.1 |
- |
4, 5 |
572 |
|
BC060867.1 |
- |
13, 14 |
2213 |
30344157 |
CR749817.1 |
- |
13, 14 |
2203 |
|
View Full Product Details