Gene Transcripts
- Gene Symbol
- CENPO
- Entrez Gene ID
- 79172
- Gene Name
- centromere protein O
- Gene Aliases
- CENP-O, ICEN-36, MCM21R
- Chromosome Location
- Chr. 2: 24793115 - 24822376 on Build GRCh38
- UniGene ID
- Hs.731569
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001199803.2 |
NP_001186732.1
|
3, 4 |
683 |
|
NM_001322101.1 |
NP_001309030.1
|
4, 5 |
797 |
|
NM_024322.3 |
NP_077298.1
|
4, 5 |
816 |
|
NR_136183.1 |
|
3, 4 |
702 |
|
GenBank mRNA |
AK027859.1 |
- |
3, 4 |
417 |
|
AK315483.1 |
- |
4, 5 |
442 |
|
AL514456.3 |
- |
3, 4 |
376 |
|
BC002870.1 |
- |
4, 5 |
473 |
3942844 |
BP366489.1 |
- |
4, 5 |
469 |
|
BX406097.2 |
- |
4, 5 |
472 |
|
CN263048.1 |
- |
3, 4 |
339 |
|
View Full Product Details