Gene Transcripts
- Gene Symbol
- PPP1R21
- Entrez Gene ID
- 129285
- Gene Name
- protein phosphatase 1 regulatory subunit 21
- Gene Aliases
- CCDC128, KLRAQ1
- Chromosome Location
- Chr. 2: 48440769 - 48515392 on Build GRCh38
- UniGene ID
- Hs.654619
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001135629.2 |
NP_001129101.1
|
6, 7 |
783 |
|
NM_001193475.1 |
NP_001180404.1
|
6, 7 |
783 |
|
NM_152994.4 |
NP_694539.1
|
6, 7 |
783 |
|
NR_024188.2 |
|
6, 7 |
822 |
|
XM_017003301.1 |
XP_016858790.1
|
6, 7 |
818 |
|
XM_017003302.1 |
XP_016858791.1
|
6, 7 |
818 |
|
GenBank mRNA |
AK122900.1 |
- |
6, 7 |
793 |
|
AK172753.1 |
- |
3, 4 |
262 |
|
AK172762.1 |
- |
6, 7 |
755 |
|
AK307755.1 |
- |
6, 7 |
706 |
|
AY092062.1 |
- |
6, 7 |
734 |
|
AY134855.1 |
- |
6, 7 |
783 |
|
BC040721.1 |
- |
6, 7 |
684 |
|
BC111059.1 |
- |
6, 7 |
725 |
5745902 |
BC117279.1 |
- |
6, 7 |
695 |
40125830 |
BC143463.1 |
- |
6, 7 |
695 |
9051971 |
BC143466.1 |
- |
6, 7 |
695 |
9051974 |
View Full Product Details