Gene Transcripts
- Gene Symbol
- YIPF7
- Entrez Gene ID
- 285525
- Gene Name
- Yip1 domain family member 7
- Gene Aliases
- FinGER9
- Chromosome Location
- Chr. 4: 44619578 - 44656868 on Build GRCh38
- UniGene ID
- Hs.596000
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK096895.1 |
- |
4 |
537 |
|
BC103995.1 |
- |
3 |
373 |
|
BC103996.1 |
- |
4 |
542 |
|
View Full Product Details