Gene Transcripts

Gene Symbol
HNRNPA3
Entrez Gene ID
220988
Gene Name
heterogeneous nuclear ribonucleoprotein A3
Gene Aliases
2610510D13Rik, D10S102, FBRNP, HNRPA3
Chromosome Location
Chr. 2: 177212694 - 177223959 on Build GRCh38
UniGene ID
Hs.516539
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_194247.2 NP_919223.1 6 817
XM_005246380.1 XP_005246437.1 6 817
XM_005246381.4 XP_005246438.1 6 723
XM_006712365.2 XP_006712428.1 6 817
XM_017003587.1 XP_016859076.1 6 723
XM_017003588.1 XP_016859077.1 6 817
GenBank mRNA AK293125.1 - 6 685
AK316434.1 - 6 685
AY363225.1 - 6 711
BC080591.1 - 2 145
BC112027.1 - 6 739 8327495
BC113470.1 - 6 739 8322522

View Full Product Details