Gene Transcripts

Gene Symbol
ZFP42
Entrez Gene ID
132625
Gene Name
ZFP42 zinc finger protein
Gene Aliases
REX-1, REX1, ZNF754, zfp-42
Chromosome Location
Chr. 4: 187994044 - 188005050 on Build GRCh38
UniGene ID
Hs.335787
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304358.1 NP_001291287.1 3 979
NM_174900.4 NP_777560.2 4 1145
XM_011531604.2 XP_011529906.1 3 1000
XM_011531605.1 XP_011529907.1 4 1150
XM_011531606.1 XP_011529908.1 4 1154
XM_011531607.2 XP_011529909.1 3 997
GenBank mRNA AF450454.1 - 1 737
AK056719.1 - 3 979
AY429593.1 - 4 1145
CN420210.1 - 1 79

View Full Product Details