Gene Transcripts

Gene Symbol
ARSK
Entrez Gene ID
153642
Gene Name
arylsulfatase family member K
Gene Aliases
TSULF
Chromosome Location
Chr. 5: 95555108 - 95605102 on Build GRCh38
UniGene ID
Hs.585051
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198150.2 NP_937793.1 7 1413
XM_005271904.3 XP_005271961.1 8 1477
GenBank mRNA AK303855.1 - 7 1465
AL832711.1 - 6 1096
AY358596.1 - 7 1373
AY875939.1 - 7 1255
BC036047.1 - 7 1275
BC050392.1 - 8 1423
BC130329.1 - 7 1351 40146360
BC144161.1 - 8 1410

View Full Product Details