Gene Transcripts

Gene Symbol
FRA10AC1
Entrez Gene ID
118924
Gene Name
fragile site, folic acid type, rare, fra(10)(q23.3) or fra(10)(q24.2) candidate 1
Gene Aliases
C10orf4, F26C11.1-like, FRA10A
Chromosome Location
Chr. 10: 93667883 - 93702572 on Build GRCh38
UniGene ID
Hs.586650
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145246.4 NP_660289.2 4 400
XM_005269515.4 XP_005269572.1 5 497
XM_005269516.4 XP_005269573.1 4 592
XM_011539259.2 XP_011537561.1 4 365
XM_011539260.2 XP_011537562.1 5 455
XM_017015656.1 XP_016871145.1 4 365
XM_017015657.1 XP_016871146.1 4 400
XM_017015658.1 XP_016871147.1 5 500
XM_017015659.1 XP_016871148.1 4 592
XM_017015660.1 XP_016871149.1 5 457
XM_017015661.1 XP_016871150.1 5 514
XM_017015662.1 XP_016871151.1 4 365
GenBank mRNA AJ431721.1 - 4 396
AJ431722.1 - 4 396
AJ431723.1 - 4 396
AJ431724.1 - 4 396
AJ431725.1 - 4 396
AK090955.1 - 4 400
AK308861.1 - 4 318
AK309016.1 - 4 513
BC018007.1 - 4 269 4278022
BM272089.1 - 4 384
BX648699.1 - 3 434
CN298116.1 - 4 379

View Full Product Details