Gene Transcripts
- Gene Symbol
- H1FOO
- Entrez Gene ID
- 132243
- Gene Name
- H1 histone family member O, oocyte specific
- Gene Aliases
- H1.8, H1oo, osH1
- Chromosome Location
- Chr. 3: 129543214 - 129555185 on Build GRCh38
- UniGene ID
- Hs.97358
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_153833.2 |
NP_722575.1
|
2, 3 |
384 |
|
GenBank mRNA |
AY158091.1 |
- |
2, 3 |
384 |
|
View Full Product Details