Gene Transcripts

Gene Symbol
FAM120AOS
Entrez Gene ID
158293
Gene Name
family with sequence similarity 120A opposite strand
Gene Aliases
C9orf10OS
Chromosome Location
Chr. 9: 93446500 - 93453592 on Build GRCh38
UniGene ID
Hs.350364
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001322224.1 NP_001309153.1 3 867
NM_198841.2 NP_942138.2 3 1796
NR_136229.1 3 1203
NR_136230.1 4 1324
NR_136231.1 4 1917
NR_136232.1 4 1131
NR_136233.1 3 954
NR_136234.1 4 988
NR_136235.1 3 1010
NR_136236.1 3 1213
NR_136237.1 4 1334
NR_136238.1 4 1075
GenBank mRNA AK056096.1 - 3 1796
AK309269.1 - 4 886
BC063650.1 - 3 760
BC080634.1 - 4 858
BC107135.1 - 4 989
BC107136.1 - 3 868
BC113074.1 - 3 868

View Full Product Details