Gene Transcripts

Gene Symbol
MRO
Entrez Gene ID
83876
Gene Name
maestro
Gene Aliases
B29, C18orf3
Chromosome Location
Chr. 18: 50795120 - 50825413 on Build GRCh38
UniGene ID
Hs.30495
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127176.1 NP_001120648.1 5 634
NM_031939.3 NP_114145.2 6 737
XM_005258365.3 XP_005258422.1 5 547
XM_017026035.1 XP_016881524.1 5 514
GenBank mRNA AB042647.1 - 6 601
AK054702.1 - 6 671
BC029860.1 - 6 624 5171860
DA416200.1 - 5 489

View Full Product Details