Gene Transcripts

Gene Symbol
HEMGN
Entrez Gene ID
55363
Gene Name
hemogen
Gene Aliases
CT155, EDAG, EDAG-1, NDR
Chromosome Location
Chr. 9: 97926791 - 97944915 on Build GRCh38
UniGene ID
Hs.176626
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018437.4 NP_060907.2 4 509
NM_197978.2 NP_932095.1 3 563
XM_005252086.1 XP_005252143.1 4 471
XM_011518845.2 XP_011517147.1 4 383
XM_011518846.2 XP_011517148.1 4 493
GenBank mRNA AF130060.2 - 3 296
AF228713.2 - 4 416
AF306682.1 - 3 345
AF306683.1 - 3 311
AF322875.1 - 4 415
AK292441.1 - 4 435
AL833094.1 - 3 356
AY244805.1 - 3 509
AY255672.1 - 3 414
BC048324.1 - 4 418 5166612
BG571634.1 - 3 387
BG717036.1 - 4 454
DA485094.1 - 3 563

View Full Product Details