Gene Transcripts

Gene Symbol
CD248
Entrez Gene ID
57124
Gene Name
CD248 molecule
Gene Aliases
CD164L1, TEM1
Chromosome Location
Chr. 11: 66314487 - 66317044 on Build GRCh38
UniGene ID
Hs.195727
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020404.2 NP_065137.1 1 2185
GenBank mRNA AF279142.1 - 1 2173
AJ295846.1 - 1 2185
AK027290.1 - 1 1307
BC051340.1 - 1 2172 6339237
BC104484.1 - 1 2185 40007855
BC105633.1 - 1 2185 40007854

View Full Product Details