Gene Transcripts
- Gene Symbol
- SH2D4A
- Entrez Gene ID
- 63898
- Gene Name
- SH2 domain containing 4A
- Gene Aliases
- PPP1R38, SH2A
- Chromosome Location
- Chr. 8: 19313443 - 19396218 on Build GRCh38
- UniGene ID
- Hs.303208
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001174159.1 |
NP_001167630.1
|
9, 10 |
1588 |
|
NM_001174160.1 |
NP_001167631.1
|
8, 9 |
1353 |
|
NM_022071.3 |
NP_071354.2
|
9, 10 |
1738 |
|
XM_011544610.1 |
XP_011542912.1
|
8, 9 |
1784 |
|
XM_017013729.1 |
XP_016869218.1
|
8, 9 |
1506 |
|
GenBank mRNA |
AK024620.1 |
- |
7, 8 |
1007 |
|
AK024799.1 |
- |
9, 10 |
1616 |
|
AK225052.1 |
- |
7, 8 |
1007 |
|
AK293301.1 |
- |
8, 9 |
1229 |
|
AY190323.1 |
- |
5, 6 |
767 |
|
BC014525.2 |
- |
6, 7 |
934 |
|
BC067117.1 |
- |
6, 7 |
947 |
|
BC082982.1 |
- |
9, 10 |
1588 |
6575224 |
View Full Product Details