Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012275.2 | NP_036407.1 | 4 | 391 | |
NM_173170.1 | NP_775262.1 | 4 | 363 | ||
GenBank mRNA | AF186094.1 | - | 4 | 392 | |
AF201830.1 | - | 3 | 257 | ||
AF230377.1 | - | 3 | 230 | ||
AJ242737.1 | - | 4 | 311 | ||
AJ242738.1 | - | 4 | 302 | ||
AK290249.1 | - | 4 | 301 | ||
AY359117.1 | - | 4 | 296 | ||
BC024747.1 | - | 4 | 363 | 4996939 |