Gene Transcripts

Gene Symbol
CLEC5A
Entrez Gene ID
23601
Gene Name
C-type lectin domain family 5 member A
Gene Aliases
CLECSF5, MDL-1, MDL1
Chromosome Location
Chr. 7: 141927356 - 141947009 on Build GRCh38
UniGene ID
Hs.446235
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301167.1 NP_001288096.1 3 351
NM_013252.2 NP_037384.1 3 327
XM_017011916.1 XP_016867405.1 3 353
XM_017011917.1 XP_016867406.1 3 353
GenBank mRNA AF139768.1 - 3 281
AJ271684.1 - 3 327
AK292766.1 - 3 316
AK295200.1 - 3 288
AK313703.1 - 3 316
BC093714.1 - 2 151 7939559
BC112099.1 - 2 151 8327567
BC113098.1 - 3 269 40076723
BC113099.1 - 3 269 40076725
BC143392.1 - 2 151

View Full Product Details