Gene Transcripts

Gene Symbol
NIPSNAP3A
Entrez Gene ID
25934
Gene Name
nipsnap homolog 3A
Gene Aliases
HSPC299, NIPSNAP4, TASSC
Chromosome Location
Chr. 9: 104747688 - 104760122 on Build GRCh38
UniGene ID
Hs.530275
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015469.1 NP_056284.1 1628
GenBank mRNA BC005935.1 - 1628

View Full Product Details