Gene Transcripts
- Gene Symbol
- SDCBP2
- Entrez Gene ID
- 27111
- Gene Name
- syndecan binding protein 2
- Gene Aliases
- SITAC, SITAC18, ST-2
- Chromosome Location
- Chr. 20: 1309909 - 1329235 on Build GRCh38
- UniGene ID
- Hs.657015
- Species
- Homo sapiens
- Gene Symbol
- FKBP1A-SDCBP2
- Entrez Gene ID
- 100528031
- Gene Name
- FKBP1A-SDCBP2 readthrough (NMD candidate)
- Gene Aliases
- Chromosome Location
- Chr. 20: 1309909 - 1393172 on Build GRCh38
- UniGene ID
- Hs.471933
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_037661.1 |
|
10 |
1421 |
|
View Full Product Details