Gene Transcripts
- Gene Symbol
- FILIP1
- Entrez Gene ID
- 27145
- Gene Name
- filamin A interacting protein 1
- Gene Aliases
- FILIP
- Chromosome Location
- Chr. 6: 75294895 - 75493829 on Build GRCh38
- UniGene ID
- Hs.696158
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001289987.1 |
NP_001276916.1
|
5, 6 |
1217 |
|
NM_001300866.1 |
NP_001287795.1
|
4, 5 |
1051 |
|
NM_015687.3 |
NP_056502.1
|
4, 5 |
1051 |
|
XM_005248713.3 |
XP_005248770.1
|
4, 5 |
1022 |
|
XM_005248715.4 |
XP_005248772.1
|
4, 5 |
1022 |
|
GenBank mRNA |
AB033101.1 |
- |
2, 3 |
439 |
|
AB086011.1 |
- |
3, 4 |
635 |
|
AK097021.1 |
- |
4, 5 |
891 |
|
AK290232.1 |
- |
4, 5 |
763 |
|
AL832009.2 |
- |
4, 5 |
851 |
|
BC136443.1 |
- |
4, 5 |
806 |
9020432 |
BC136444.1 |
- |
5, 6 |
972 |
9020433 |
BX647178.1 |
- |
4, 5 |
1002 |
|
View Full Product Details