Gene Transcripts

Gene Symbol
TAS2R13
Entrez Gene ID
50838
Gene Name
taste 2 receptor member 13
Gene Aliases
T2R13, TRB3
Chromosome Location
Chr. 12: 10907926 - 10909562 on Build GRCh38
UniGene ID
Hs.679407
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023920.2 NP_076409.1 1 392
GenBank mRNA BC069376.1 - 1 128
BC095518.1 - 1 392 30706594
BC115402.1 - 1 121

View Full Product Details