Gene Transcripts

Gene Symbol
IFI44
Entrez Gene ID
10561
Gene Name
interferon induced protein 44
Gene Aliases
MTAP44, TLDC5, p44
Chromosome Location
Chr. 1: 78648554 - 78664078 on Build GRCh38
UniGene ID
Hs.82316
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006417.4 NP_006408.3 5 948
NR_135640.1 4 484
NR_135641.1 5 951
XM_005270380.3 XP_005270437.1 5 2186
XM_011540516.2 XP_011538818.1 5 2186
XM_017000083.1 XP_016855572.1 5 2186
GenBank mRNA AK302527.1 - 5 909
AK316278.1 - 4 441
AK316476.1 - 5 906
BC022870.1 - 5 913 4071404

View Full Product Details