Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007361.3 | NP_031387.3 | 8 | 2170 | |
XM_005267405.4 | XP_005267462.1 | 7 | 2254 | ||
XM_005267406.4 | XP_005267463.1 | 7 | 2254 | ||
XM_005267407.4 | XP_005267464.1 | 8 | 2173 | ||
GenBank mRNA | AJ223500.1 | - | 8 | 1936 | |
AK291652.1 | - | 8 | 1998 | ||
AK300462.1 | - | 9 | 1808 | ||
AU119925.1 | - | 1 | 117 | ||
BC035608.1 | - | 4 | 772 | 5205293 | |
BX648241.1 | - | 9 | 1943 | ||
D86425.1 | - | 8 | 1939 |