Gene Transcripts
- Gene Symbol
- HABP4
- Entrez Gene ID
- 22927
- Gene Name
- hyaluronan binding protein 4
- Gene Aliases
- IHABP-4, IHABP4, Ki-1/57, SERBP1L
- Chromosome Location
- Chr. 9: 96450155 - 96491336 on Build GRCh38
- UniGene ID
- Hs.494567
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF241831.1 |
- |
8 |
1396 |
|
U77327.1 |
- |
7 |
979 |
|
View Full Product Details