Gene Transcripts

Gene Symbol
GGA2
Entrez Gene ID
23062
Gene Name
golgi associated, gamma adaptin ear containing, ARF binding protein 2
Gene Aliases
VEAR
Chromosome Location
Chr. 16: 23463542 - 23510494 on Build GRCh38
UniGene ID
Hs.460336
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015044.4 NP_055859.1 17 1870
XM_017023074.1 XP_016878563.1 18 5559
XM_017023075.1 XP_016878564.1 17 1873
GenBank mRNA AB029003.1 - 14 1500
AF165531.1 - 17 1808
AF190863.1 - 17 1814
AF233522.1 - 17 1813
AF323754.1 - 18 1726
AK024619.1 - 17 1869
AK093769.1 - 2 753
BC000284.1 - 17 1852 3356490

View Full Product Details