Gene Transcripts

Gene Symbol
MYOM1
Entrez Gene ID
8736
Gene Name
myomesin 1
Gene Aliases
SKELEMIN
Chromosome Location
Chr. 18: 3066807 - 3220108 on Build GRCh38
UniGene ID
Hs.464469
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK125322.1 - 4 871
BC116183.1 - 4 612 40073728
X69090.1 - 2 246

View Full Product Details