Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003827.3 | NP_003818.2 | 11 | 1616 | |
NR_038456.1 | 12 | 1272 | |||
NR_038457.1 | 9 | 1419 | |||
XM_011527436.1 | XP_011525738.1 | 11 | 1290 | ||
XM_011527437.1 | XP_011525739.1 | 10 | 1369 | ||
GenBank mRNA | AK098401.1 | - | 11 | 1449 | |
AK123456.1 | - | 1 | 1641 | ||
AK126519.1 | - | 5 | 3658 | ||
BC001165.1 | - | 11 | 1388 | 3533668 | |
BC007432.2 | - | 11 | 1379 | 2962847 | |
BC028234.1 | - | 11 | 1434 | 5245770 | |
BC091511.1 | - | 11 | 1395 | 30512911 | |
U39412.1 | - | 12 | 1022 |