Gene Transcripts

Gene Symbol
SNX4
Entrez Gene ID
8723
Gene Name
sorting nexin 4
Gene Aliases
ATG24B
Chromosome Location
Chr. 3: 125446644 - 125520214 on Build GRCh38
UniGene ID
Hs.507243
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003794.3 NP_003785.1 12 1178
NR_073435.1 11 1056
XM_017007414.1 XP_016862903.1 13 1266
GenBank mRNA AF065485.1 - 13 1136
AF130078.1 - 7 545
AK001835.1 - 12 1140
AK298972.1 - 11 1023
BC018762.1 - 12 1154 4641014

View Full Product Details