Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_011537317.2 | XP_011535619.1 | 5 | 707 | |
XM_011537318.2 | XP_011535620.1 | 5 | 707 | ||
XM_017021760.1 | XP_016877249.1 | 5 | 484 | ||
GenBank mRNA | AK000913.1 | - | 5 | 438 | |
AK293236.1 | - | 4 | 322 | ||
AK295010.1 | - | 5 | 446 | ||
AK304695.1 | - | 5 | 443 | ||
BC006001.1 | - | 4 | 324 | ||
BC056687.1 | - | 5 | 454 | ||
BM835709.1 | - | 5 | 483 |