Gene Transcripts

Gene Symbol
RHOBTB1
Entrez Gene ID
9886
Gene Name
Rho related BTB domain containing 1
Gene Aliases
Chromosome Location
Chr. 10: 60869438 - 61001486 on Build GRCh38
UniGene ID
Hs.737374
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242359.1 NP_001229288.1 7 1012
NM_014836.4 NP_055651.1 6 940
NR_024554.2 7 814
NR_024555.1 7 778
XM_006718084.1 XP_006718147.1 6 748
XM_006718085.1 XP_006718148.1 6 775
XM_011540424.1 XP_011538726.1 7 806
XM_011540425.1 XP_011538727.1 7 806
XM_011540426.1 XP_011538728.1 7 806
XM_011540428.1 XP_011538730.1 7 806
XM_017017000.1 XP_016872489.1 6 995
XM_017017001.1 XP_016872490.1 6 991
XM_017017002.1 XP_016872491.1 6 991
XM_017017003.1 XP_016872492.1 8 881
XM_017017004.1 XP_016872493.1 8 883
XM_017017005.1 XP_016872494.1 7 788
XM_017017006.1 XP_016872495.1 7 837
XM_017017007.1 XP_016872496.1 5 887
XM_017017008.1 XP_016872497.1 7 1048
GenBank mRNA AB018283.3 - 6 831
AK075129.1 - 7 778
AK307630.1 - 5 510
BC032848.1 - 7 989 4830471
BC041791.1 - 6 731 5297756
CF593541.1 - 6 755

View Full Product Details