Gene Transcripts
- Gene Symbol
- RHOBTB1
- Entrez Gene ID
- 9886
- Gene Name
- Rho related BTB domain containing 1
- Gene Aliases
- Chromosome Location
- Chr. 10: 60869438 - 61001486 on Build GRCh38
- UniGene ID
- Hs.737374
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001242359.1 |
NP_001229288.1
|
7, 8 |
1904 |
|
NM_014836.4 |
NP_055651.1
|
6, 7 |
1832 |
|
NR_024554.2 |
|
7, 8 |
1706 |
|
NR_024555.1 |
|
7, 8 |
1670 |
|
XM_006718084.1 |
XP_006718147.1
|
6, 7 |
1640 |
|
XM_006718085.1 |
XP_006718148.1
|
6, 7 |
1667 |
|
XM_011540424.1 |
XP_011538726.1
|
7, 8 |
1698 |
|
XM_011540425.1 |
XP_011538727.1
|
7, 8 |
1698 |
|
XM_011540426.1 |
XP_011538728.1
|
7, 8 |
1698 |
|
XM_011540428.1 |
XP_011538730.1
|
7, 8 |
1698 |
|
XM_017017000.1 |
XP_016872489.1
|
6, 7 |
1887 |
|
XM_017017001.1 |
XP_016872490.1
|
6, 7 |
1883 |
|
XM_017017002.1 |
XP_016872491.1
|
6, 7 |
1883 |
|
XM_017017003.1 |
XP_016872492.1
|
8, 9 |
1773 |
|
XM_017017004.1 |
XP_016872493.1
|
8, 9 |
1775 |
|
XM_017017005.1 |
XP_016872494.1
|
7, 8 |
1680 |
|
XM_017017006.1 |
XP_016872495.1
|
7, 8 |
1729 |
|
XM_017017007.1 |
XP_016872496.1
|
5, 6 |
1779 |
|
XM_017017008.1 |
XP_016872497.1
|
7, 8 |
1940 |
|
GenBank mRNA |
AB018283.3 |
- |
6, 7 |
1723 |
|
AK075129.1 |
- |
7, 8 |
1670 |
|
AK307630.1 |
- |
5, 6 |
1402 |
|
BC032848.1 |
- |
7, 8 |
1881 |
4830471 |
BC041791.1 |
- |
6, 7 |
1623 |
5297756 |
DA830304.1 |
- |
1, 2 |
578 |
|
View Full Product Details