Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001286102.1 | NP_001273031.1 | 2 | 247 | |
NM_003269.4 | NP_003260.1 | 2 | 840 | ||
GenBank mRNA | AB307709.1 | - | 2 | 132 | |
AF220532.1 | - | 2 | 839 | ||
AF411525.1 | - | 2 | 373 | ||
AK131541.1 | - | 2 | 247 | ||
BC028031.1 | - | 2 | 337 | 5242079 | |
HQ692846.1 | - | 2 | 132 | ||
Y13276.1 | - | 2 | 300 |