Gene Transcripts

Gene Symbol
USP1
Entrez Gene ID
7398
Gene Name
ubiquitin specific peptidase 1
Gene Aliases
UBP
Chromosome Location
Chr. 1: 62436304 - 62451804 on Build GRCh38
UniGene ID
Hs.35086
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017415.1 NP_001017415.1 7 1630
NM_001017416.1 NP_001017416.1 7 1535
NM_003368.4 NP_003359.3 7 2060
GenBank mRNA AB014458.1 - 7 1411
AB208893.1 - 7 1691
AF117386.1 - 7 1547
AK312423.1 - 7 1723
AL117575.1 - 6 1382
BC023216.1 - 3 895
BC050525.1 - 7 1764 6473568

View Full Product Details