Gene Transcripts
- Gene Symbol
- MBD3
- Entrez Gene ID
- 53615
- Gene Name
- methyl-CpG binding domain protein 3
- Gene Aliases
- Chromosome Location
- Chr. 19: 1576671 - 1592761 on Build GRCh38
- UniGene ID
- Hs.178728
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001281453.1 |
NP_001268382.1
|
5, 6 |
807 |
|
NM_001281454.1 |
NP_001268383.1
|
5, 6 |
711 |
|
GenBank mRNA |
AF072247.1 |
- |
5, 6 |
700 |
|
AK001474.1 |
- |
4, 5 |
507 |
|
AK021532.1 |
- |
4, 5 |
461 |
|
AK290882.1 |
- |
5, 6 |
754 |
|
BC009372.1 |
- |
5, 6 |
643 |
4122128 |
BC009438.1 |
- |
3, 4 |
326 |
|
BC032443.1 |
- |
3, 4 |
315 |
|
BC043619.1 |
- |
5, 6 |
661 |
6156267 |
View Full Product Details