Gene Transcripts

Gene Symbol
KDM6A
Entrez Gene ID
7403
Gene Name
lysine demethylase 6A
Gene Aliases
KABUK2, UTX, bA386N14.2
Chromosome Location
Chr. X: 44873175 - 45112612 on Build GRCh38
UniGene ID
Hs.522616
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291415.1 NP_001278344.1 12, 13 1579
NM_021140.3 NP_066963.2 12, 13 1579
XM_005272656.4 XP_005272713.1 12, 13 1579
XM_005272659.4 XP_005272716.1 12, 13 1579
XM_011543957.2 XP_011542259.1 13, 14 1636
XM_011543958.2 XP_011542260.1 12, 13 1579
XM_011543959.2 XP_011542261.1 13, 14 1636
XM_011543960.2 XP_011542262.1 13, 14 1636
XM_011543962.2 XP_011542264.1 13, 14 1636
XM_011543964.2 XP_011542266.1 12, 13 1579
XM_011543966.2 XP_011542268.1 13, 14 1636
XM_011543967.2 XP_011542269.1 13, 14 1636
XM_011543970.2 XP_011542272.1 12, 13 1579
XM_011543974.1 XP_011542276.1 12, 13 1579
XM_011543975.2 XP_011542277.1 8, 9 749
GenBank mRNA AB208795.1 - 12, 13 1532
AF000992.1 - 12, 13 1228
AF000993.1 - 12, 13 1228
AK304114.1 - 12, 13 1615
AK307691.1 - 2, 3 314
BC093868.1 - 12, 13 1225 7939713
BC113381.1 - 12, 13 1225 8322433
BC143277.1 - 12, 13 1225 9051779

Gene Symbol
UTY
Entrez Gene ID
7404
Gene Name
ubiquitously transcribed tetratricopeptide repeat containing, Y-linked
Gene Aliases
KDM6AL, UTY1
Chromosome Location
Chr. Y: 13230770 - 13480670 on Build GRCh38
UniGene ID
Hs.115277
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001258249.1 NP_001245178.1 12, 13 2198
NM_001258252.1 NP_001245181.1 12, 13 2198
NM_001258254.1 NP_001245183.1 12, 13 2198
NM_001258255.1 NP_001245184.1 12, 13 2198
NM_001258256.1 NP_001245185.1 12, 13 2198
NM_001258261.1 NP_001245190.1 11, 12 2099
NM_001258262.1 NP_001245191.1 12, 13 2198
NM_001258264.1 NP_001245193.1 12, 13 2150
NM_001258266.1 NP_001245195.1 12, 13 2198
NM_001258268.1 NP_001245197.1 12, 13 2198
NM_001258269.1 NP_001245198.1 12, 13 2198
NR_047598.1 13, 14 2299
NR_047600.1 11, 12 2163
NR_047601.1 12, 13 2198
NR_047607.1 12, 13 2198
NR_047608.1 12, 13 2150
NR_047611.1 12, 13 2198
NR_047612.1 12, 13 2198
NR_047613.1 11, 12 2148
NR_047618.1 12, 13 2198
NR_047620.1 12, 13 2198
NR_047621.1 12, 13 2269
NR_047622.1 11, 12 2099
NR_047623.1 12, 13 2198
NR_047633.1 12, 13 2198
NR_047643.1 12, 13 2198
NR_047644.1 12, 13 2198
NR_047645.1 12, 13 2269
XM_005262518.3 XP_005262575.1 12, 13 1544
XM_006724875.3 XP_006724938.1 12, 13 1544
XM_011531441.2 XP_011529743.1 12, 13 1544
XM_011531442.2 XP_011529744.1 12, 13 1544
XM_011531443.2 XP_011529745.1 12, 13 1544
XM_011531446.2 XP_011529748.1 12, 13 1544
XM_011531449.2 XP_011529751.1 12, 13 1544
XM_011531453.2 XP_011529755.1 12, 13 1544
XM_011531454.2 XP_011529756.1 12, 13 1544
XM_011531455.2 XP_011529757.1 12, 13 1544
XM_011531457.2 XP_011529759.1 12, 13 1544
XM_011531458.2 XP_011529760.1 12, 13 1544
XM_011531459.2 XP_011529761.1 12, 13 1544
XM_011531460.2 XP_011529762.1 12, 13 1544
XM_011531461.2 XP_011529763.1 12, 13 1544
XM_011531462.2 XP_011529764.1 12, 13 1544
XM_011531463.2 XP_011529765.1 12, 13 1544
XM_011531464.2 XP_011529766.1 12, 13 1544
XM_017030066.1 XP_016885555.1 12, 13 1544
XM_017030067.1 XP_016885556.1 12, 13 1544
XM_017030070.1 XP_016885559.1 12, 13 1544
XM_017030071.1 XP_016885560.1 12, 13 1544
XM_017030074.1 XP_016885563.1 12, 13 1544
GenBank mRNA AY769061.1 - 13, 14 1294
AY769066.1 - 12, 13 1193
CR936684.1 - 1, 2 29
DQ140389.1 - 12, 13 1193
DQ140390.1 - 12, 13 1193
DQ345542.1 - 11, 12 1158
DQ345544.1 - 12, 13 1193
DQ350456.1 - 11, 12 1158
EF491796.1 - 1, 2 113
EF612660.1 - 12, 13 1193
EF612662.1 - 12, 13 1193
EF612663.1 - 12, 13 1193
EF612668.1 - 12, 13 1193
EF612675.1 - 12, 13 1145
EF612677.1 - 12, 13 1193
EF612679.1 - 12, 13 1193
EF612683.1 - 11, 12 1094
EF612684.1 - 12, 13 1193
EF612685.1 - 12, 13 1193
EF612686.1 - 12, 13 1193
EF612689.1 - 12, 13 1193
EF612691.1 - 12, 13 1193
EF612695.1 - 12, 13 1145
EF653168.1 - 12, 13 1193
EF653175.1 - 13, 14 1299
EF653177.1 - 12, 13 1193
EF653200.1 - 12, 13 1193
EF653207.1 - 12, 13 1193
EF653216.1 - 11, 12 1110
EF653219.1 - 12, 13 1193
EF653220.1 - 12, 13 1193
EF653222.1 - 13, 14 1273
EF653224.1 - 12, 13 1193
EF685285.1 - 12, 13 1145
EF685286.1 - 12, 13 1193
EF685288.1 - 12, 13 1193
EF685290.1 - 11, 12 1143
EF685295.1 - 12, 13 1193
EF685297.1 - 12, 13 1193
EF685298.1 - 12, 13 1264
EF685300.1 - 11, 12 1094
EF685301.1 - 12, 13 1193
EF685306.1 - 11, 12 1094
EF685330.1 - 11, 12 1094
EU026280.1 - 12, 13 1216
EU026290.1 - 10, 11 1103
EU026292.1 - 12, 13 1193
EU026302.1 - 13, 14 1322
EU026303.1 - 12, 13 1188
EU026309.1 - 12, 13 1145
EU026311.1 - 12, 13 1193
EU026314.1 - 13, 14 1299
EU026317.1 - 12, 13 1193
EU026320.1 - 13, 14 1299
EU026321.1 - 12, 13 1188
EU026323.1 - 11, 12 1153
EU026325.1 - 12, 13 1193
EU026335.1 - 12, 13 1193
EU026342.1 - 10, 11 1108
EU116343.1 - 6, 7 586
EU116347.1 - 10, 11 1064

View Full Product Details