Gene Transcripts
- Gene Symbol
- LUC7L3
- Entrez Gene ID
- 51747
- Gene Name
- LUC7 like 3 pre-mRNA splicing factor
- Gene Aliases
- CRA, CREAP-1, CROP, LUC7A, OA48-18, hLuc7A
- Chromosome Location
- Chr. 17: 50719565 - 50754269 on Build GRCh38
- UniGene ID
- Hs.130293
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_006107.3 |
NP_006098.2
|
2, 3 |
330 |
|
NM_016424.4 |
NP_057508.2
|
2, 3 |
330 |
|
XM_005257448.1 |
XP_005257505.1
|
2, 3 |
298 |
|
XM_005257449.2 |
XP_005257506.1
|
2, 3 |
298 |
|
XM_005257452.1 |
XP_005257509.1
|
2, 3 |
298 |
|
GenBank mRNA |
AK001925.1 |
- |
2, 3 |
264 |
|
AK023672.1 |
- |
2, 3 |
292 |
|
AK225980.1 |
- |
2, 3 |
298 |
|
AK290540.1 |
- |
2, 3 |
297 |
|
BC002925.2 |
- |
2, 3 |
271 |
|
BC047043.1 |
- |
2, 3 |
279 |
|
DA231851.1 |
- |
2, 3 |
330 |
|
DQ013876.1 |
- |
2, 3 |
266 |
|
View Full Product Details