Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001163034.1 | NP_001156506.1 | 3 | 1115 | |
NM_020761.2 | NP_065812.1 | 3 | 1115 | ||
GenBank mRNA | AB082951.1 | - | 3 | 310 | |
AK055912.1 | - | 3 | 961 | ||
AK296321.1 | - | 3 | 1110 | ||
AY090663.1 | - | 3 | 672 | ||
BC064515.1 | - | 3 | 1063 | ||
BC136652.1 | - | 3 | 505 | 9020642 | |
BC136654.1 | - | 3 | 505 | 9020644 |