Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000953.2 | NP_000944.1 | 1 | 652 | |
NM_001281469.1 | NP_001268398.1 | 1 | 652 | ||
XM_005267891.3 | XP_005267948.1 | 1 | 1109 | ||
GenBank mRNA | BC040968.1 | - | 1 | 652 | |
CD013871.1 | - | 1 | 553 | ||
EF577397.1 | - | 1 | 550 |