Gene Transcripts

Gene Symbol
RPS15
Entrez Gene ID
6209
Gene Name
ribosomal protein S15
Gene Aliases
RIG, S15
Chromosome Location
Chr. 19: 1438364 - 1440497 on Build GRCh38
UniGene ID
Hs.406683
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001018.4 NP_001009.1 4 435
NM_001308226.1 NP_001295155.1 4 526
GenBank mRNA BC064908.1 - 1 379
BC105810.1 - 4 413 6744553
BC141832.1 - 1 379 40148744
BE798984.1 - 4 614
BG253776.1 - 1 374
BG705752.1 - 4 532
BM558707.1 - 4 514
BU602036.1 - 4 436
J02984.1 - 4 402

View Full Product Details