Gene Transcripts
- Gene Symbol
- PMAIP1
- Entrez Gene ID
- 5366
- Gene Name
- phorbol-12-myristate-13-acetate-induced protein 1
- Gene Aliases
- APR, NOXA
- Chromosome Location
- Chr. 18: 59899960 - 59904306 on Build GRCh38
- UniGene ID
- Hs.96
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_021127.2 |
NP_066950.1
|
2 |
1841 |
|
GenBank mRNA |
D90070.1 |
- |
2 |
1791 |
|
View Full Product Details