Gene Transcripts
- Gene Symbol
- IL3
- Entrez Gene ID
- 3562
- Gene Name
- interleukin 3
- Gene Aliases
- IL-3, MCGF, MULTI-CSF
- Chromosome Location
- Chr. 5: 132060654 - 132063203 on Build GRCh38
- UniGene ID
- Hs.694
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000588.3 |
NP_000579.2
|
4, 5 |
396 |
|
GenBank mRNA |
BC066272.1 |
- |
4, 5 |
389 |
|
BC066273.1 |
- |
4, 5 |
389 |
6971774 |
BC066274.1 |
- |
4, 5 |
389 |
6971775 |
BC066275.1 |
- |
4, 5 |
389 |
6971776 |
BC066276.1 |
- |
4, 5 |
389 |
6971777 |
BC069472.1 |
- |
4, 5 |
392 |
7262122 |
M14743.1 |
- |
4, 5 |
352 |
|
M17115.1 |
- |
4, 5 |
381 |
|
M20137.1 |
- |
4, 5 |
396 |
|
View Full Product Details