Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005288.3 | NP_005279.1 | 2 | 1033 | |
XM_005266360.3 | XP_005266417.1 | 2 | 630 | ||
GenBank mRNA | AK290220.1 | - | 2 | 1034 | |
AK294378.1 | - | 2 | 489 | ||
AK307756.1 | - | 2 | 1032 | ||
AK312602.1 | - | 2 | 889 | ||
BC067448.1 | - | 1 | 811 | ||
BC067449.1 | - | 1 | 811 | ||
BC067450.1 | - | 1 | 811 | ||
BC067451.1 | - | 1 | 811 | ||
BC067452.1 | - | 1 | 811 | ||
BC112144.1 | - | 1 | 886 | 8327612 | |
BC112146.1 | - | 1 | 886 | 8327614 | |
BM727022.1 | - | 1 | 145 | ||
HQ995525.1 | - | 1 | 811 |