Gene Transcripts

Gene Symbol
CSN1S1
Entrez Gene ID
1446
Gene Name
casein alpha s1
Gene Aliases
CASA, CSN1
Chromosome Location
Chr. 4: 69931062 - 69946570 on Build GRCh38
UniGene ID
Hs.3155
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025104.1 NP_001020275.1 4, 5 155
NM_001890.1 NP_001881.1 4, 5 155
XM_006714089.2 XP_006714152.1 4, 5 174
XM_006714090.2 XP_006714153.1 4, 5 174
XM_006714091.2 XP_006714154.1 4, 5 174
GenBank mRNA BC128227.1 - 4, 5 144
BC128228.1 - 4, 5 144 40114622
BG217918.1 - 3, 4 117
DQ064604.1 - 3, 4 106
U23157.1 - 4, 5 134
X78416.1 - 4, 5 155
X98084.1 - 3, 4 83

View Full Product Details