Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001938.2 | NP_001929.1 | 3 | 1521 | |
GenBank mRNA | AK057541.1 | - | 4 | 1383 | |
AK311288.1 | - | 3 | 679 | ||
AL137673.1 | - | 2 | 1213 | ||
AL833729.1 | - | 3 | 1504 | ||
BC002809.2 | - | 3 | 1498 | 3634908 | |
BC035507.2 | - | 3 | 1521 | 4555131 | |
BC068553.1 | - | 3 | 1520 | 4822070 |